Learn More
Abcam Phomo x anthone A, apoptotic agent, 1MG
Catalog No. 502705883
Encompass_Preferred
MW 750.7 Da, Purity >97%. Cancer cell selective apoptotic agent. Antibiotic agent. Induces apoptosis in cancer cells lines with 100 fold greater potency than noncancerous lines. Shows immunostimulant effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5883 Supplier Abcam Supplier No. AB1443261MG
MW 750.7 Da, Purity >97%. Cancer cell selective apoptotic agent. Antibiotic agent. Induces apoptosis in cancer cells lines with 100 fold greater potency than noncancerous lines. Shows immunostimulant effects.
The product is subject to the following: Abcam Restricted Use Statement