Learn More
TARGETMOL CHEMICALS INC PHORBOL 12-MYRISTATE 13- 10MG
SDP
Catalog No. 502576153
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Phorbol 12-myristate 13-acetate (PMA) (PMA) is a dual SphK and PKC activator. purity: 99%
Catalog No. 50-257-6153 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ019810MG
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Phorbol 12-myristate 13-acetate (PMA) (PMA) is a dual SphK and PKC activator. purity: 99%