Learn More
BIOGEMS INTERNATIONAL INC Phorbol 12-myristate 13-acetate 5mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 16529815MG
Encompass
Phorbol 12-myristate 13-acetate (PMA) is a diester of phorbol and activates the signal transduction enzyme protein kinase C (PKC). It is used as a tumor promoter and to stimulate division of B-cells. With ionomycin, PMA induces the activation of many cell types to produce cytokines.
Catalog No. 50-152-9802