Learn More
Selleck Chemical LLC Phorbol 12-myristate 13-acetat
SDP
Catalog No. 502173307
Encompass_Preferred
Phorbol 12-myristate 13-acetate (PMA,12-O-Tetradecanoylphorbol-13-acetate, TPA, Phorbol myristate acetate), a potent activator of PKC, is active at nanomolar concentrations. Phorbol 12-myristate 13-acetate (PMA) induces sphingosine-1-phosphate (S1P). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3307 Supplier Selleck Chemical LLC Supplier No. S77915MG
Phorbol 12-myristate 13-acetate (PMA,12-O-Tetradecanoylphorbol-13-acetate, TPA, Phorbol myristate acetate), a potent activator of PKC, is active at nanomolar concentrations. Phorbol 12-myristate 13-acetate (PMA) induces sphingosine-1-phosphate (S1P). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.