Learn More
Sigma Aldrich Fine Chemicals Biosciences Phorbol 12-myristate 13-acetate, 16561-29-8, MFCD00036736, 5mg
Catalog No. 501656917
Encompass
Empirical Formula (Hill Notation) C36H56O8, ≥99% (TLC), Molecular Weight 616.83, film or powder, Synonym: 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA; Phorbol 12-myristate 13-acetate is a phorbol ester. It works as a tumor promoter and is involved in gene transcription, cell growth, differentiation, programmed cell death, immune pathway and receptor desensitization via protein kinase C (PKC) signaling pathways.
Catalog No. 50-165-6917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P81395MG
Empirical Formula (Hill Notation) C36H56O8, ≥99% (TLC), Molecular Weight 616.83, film or powder, Synonym: 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA; Phorbol 12-myristate 13-acetate is a phorbol ester. It works as a tumor promoter and is involved in gene transcription, cell growth, differentiation, programmed cell death, immune pathway and receptor desensitization via protein kinase C (PKC) signaling pathways.