Learn More
Abcam Phorbol 12-myristate 13-acetate (PMA), PKC activator, 1MG
Supplier:  Abcam AB1202971MG
Encompass_Preferred
Phorbol 12-myristate 13-acetate (PMA), PKC activator is a potent nanomolar activator of protein kinase C in vivo and in vitro. Binds to C1 domain of protein kinase C, induces membrane translocation and enzyme activation. Also reported to have actions on non-kinase proteins including chimaerins, RasGRP and Unc-13/Munc-13. Extremely potent mouse skin tumor promoter. - Cited in over 45 publications - Purity > 99% - Supplied in DMSO (10mM)
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0840361