Learn More
Medchemexpress LLC Phorbol 12-myristate 13-acetate | 16561-29-8 | 99.8% | 616.83 | 100 MG
SDP
Catalog No. 5000388210
Encompass_Preferred
Phorbol 12-myristate 13-acetate (PMA), also known as TPA or Phorbol myristate acetate, is a phorbol ester. It acts as a dual SphK and protein kinase C (PKC) activator, and also functions as a NF-κB activator. PMA induces differentiation in THP-1 cells and is known to induce a pronounced inflammatory response mediated by protein kinase C (PKC), specifically by activating PLA2.
  • Dual SphK and protein kinase C activator
  • NF-κB activator
  • Induces differentiation in THP-1 cells
  • IC₅₀ for PKC is 11.7 nM (EC50)
  • Inhibits endothelial cell migration through activating the PKC-δ/Syk/NF-κB-mediated up-regulation of Thy-1
Catalog No. 50-003-88210 Supplier Medchemexpress LLC Supplier No. HY18739100MG
Phorbol 12-myristate 13-acetate (PMA), also known as TPA or Phorbol myristate acetate, is a phorbol ester. It acts as a dual SphK and protein kinase C (PKC) activator, and also functions as a NF-κB activator. PMA induces differentiation in THP-1 cells and is known to induce a pronounced inflammatory response mediated by protein kinase C (PKC), specifically by activating PLA2.
  • Dual SphK and protein kinase C activator
  • NF-κB activator
  • Induces differentiation in THP-1 cells
  • IC₅₀ for PKC is 11.7 nM (EC50)
  • Inhibits endothelial cell migration through activating the PKC-δ/Syk/NF-κB-mediated up-regulation of Thy-1