Learn More
Medchemexpress LLC Phorbol 12-myristate 13-acetate | 16561-29-8 | 99.8% | 616.83 | 25 MG
SDP
Catalog No. 5000384479
Encompass_Preferred
Phorbol 12-myristate 13-acetate (PMA) is a phorbol ester and a dual SphK and protein kinase C (PKC) activator. It also acts as an NF-κB activator and induces differentiation in THP-1 cells. This product is intended for research and analytical applications, and extreme caution is advised when handling due to its potent tumor-promoting properties.
  • Functions as a dual SphK and protein kinase C (PKC) activator.
  • Acts as an NF-κB activator.
  • Induces differentiation in THP-1 cells.
  • Suitable for research and analytical applications.
  • Can be used to create models of ear edema in vivo.
Catalog No. 50-003-84479 Supplier Medchemexpress LLC Supplier No. 25MGHY18739
Phorbol 12-myristate 13-acetate (PMA) is a phorbol ester and a dual SphK and protein kinase C (PKC) activator. It also acts as an NF-κB activator and induces differentiation in THP-1 cells. This product is intended for research and analytical applications, and extreme caution is advised when handling due to its potent tumor-promoting properties.
  • Functions as a dual SphK and protein kinase C (PKC) activator.
  • Acts as an NF-κB activator.
  • Induces differentiation in THP-1 cells.
  • Suitable for research and analytical applications.
  • Can be used to create models of ear edema in vivo.