Learn More
Medchemexpress LLC Phorbol 12-myristate 13-acetate | 16561-29-8 | 99.8% | 616.83 | 50 MG
SDP
Catalog No. 5000387871
Encompass_Preferred
Phorbol 12-myristate 13-acetate (PMA) is a phorbol ester that acts as a dual SphK and protein kinase C (PKC) activator. It also functions as an NF-κB activator and induces differentiation in THP-1 cells. This product is for research use only and has been validated by MedChemExpress to induce differentiation in THP-1 cells.
  • Dual SphK and protein kinase C (PKC) activator
  • NF-κB activator
  • Induces differentiation in THP-1 cells
  • For research use only
Catalog No. 50-003-87871 Supplier Medchemexpress LLC Supplier No. HY1873950MG
Phorbol 12-myristate 13-acetate (PMA) is a phorbol ester that acts as a dual SphK and protein kinase C (PKC) activator. It also functions as an NF-κB activator and induces differentiation in THP-1 cells. This product is for research use only and has been validated by MedChemExpress to induce differentiation in THP-1 cells.
  • Dual SphK and protein kinase C (PKC) activator
  • NF-κB activator
  • Induces differentiation in THP-1 cells
  • For research use only