Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phorbol 12-myristate 13-acetate, >99.5%, For HPLC analysis, MP Biomedicals™

Catalog No. 0215186405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-151-86405 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-151-86405 Supplier MP Biomedicals, Inc Supplier No. 0215186405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 16561-29-8
Color Colorless
Assay Percent Range >99%
Molecular Formula C36H56O8
Quantity 5 mg
MDL Number MFCD00036736
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Weight (g/mol) 616.84
PubChem CID 27924
ChEBI CHEBI:37537
Formula Weight 616.84
Percent Purity 0.995
Chemical Name or Material Phorbol 12-myristate 13-acetate, For HPLC analysis
Show More Show Less
Recommended Storage Store at -20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.