Learn More
Invivogen PMA, 5 mg
SDP
Supplier:  Invivogen TLRLPMA
Encompass
Phorbol 12-myristate 13-acetate (PMA), also known as 12-O-tetradecanoylphorbol 13-acetate (TPA), is a specific activator of Protein Kinase C (PKC) and hence activates nuclear factor-kappa B (NF-KB). NF-κB is a transcription factor that regulates numerous physiological functions and is involved in the pathogenesis of various diseases. It has been identified as a potential therapeutic target in inflammatory processes, cancer, and autoimmune diseases.
Catalog No. NC9619504