Learn More
LAB ALLEY Phosphoric Acid 30% Solution Food Grade (FCC) Kosher Halal
SDP
Catalog No. NC3584917
Encompass
Phosphoric acid, also known as orthophosphoric acid, is a weak triprotic acid. At 30% it is a colorless and odorless liquid. The FCC Food Grade designation ensures that this product complies with the purity standards set by the United States Pharmacopeia and is therefore suitable for food use. Kosher certification requires that all ingredients equipment and processing comply with the Jewish dietary law.
Catalog No. NC3584917 Supplier LAB ALLEY Supplier No. PHAF301GAL
Phosphoric acid, also known as orthophosphoric acid, is a weak triprotic acid. At 30% it is a colorless and odorless liquid. The FCC Food Grade designation ensures that this product complies with the purity standards set by the United States Pharmacopeia and is therefore suitable for food use. Kosher certification requires that all ingredients equipment and processing comply with the Jewish dietary law.