Learn More
FUJIFILM BIOSCIENCES INC PHOS-TAG ACRYLAMIDE 10 MG
Supplier:  FUJIFILM BIOSCIENCES INC 30493521
Encompass_Preferred
*Orders for this supplier are non-cancellable/non-returnable once they are processed.* Phos-taga, c acrylamide is an acrylamide-pendant Phos-taga, c molecule. Phos-taga, c SDS-PaGE, where a polyacrylamide gel containing Phos-taga, c is used, can be prepared by adding Phos-taga, c acrylamide and MnCl2 in Resolving Gel when preparing SDS-PaGE gels. During migration, the phosphorylated proteins with the phosphate group bound to the divalent metal ions in Phos-taga, c. This decreases the migration speed and phosphorylaed/non-phosphorylated proteins are separated. after separation, the gel can be utilized for western blotting and mass spectrometry. Various Phos-taga, c SDS-PaGE precast gels are also available. Please see 8¿4¿SuperSep Phos-taga, c on the page #19 for the details.
Catalog No. NC0118170