Learn More
Apexbio Technology LLC Phos binding reagent acrylamide,5mg Cas No: NA MFCD: NA
SDP
Supplier:  Apexbio Technology LLC F40025
Encompass_Preferred
Phos binding reagents are specific and selective phosphate-binding reagents, and exhibit preferential ionic interactions with phosphorylated ions on phosphorylated proteins or peptides at neutral pH. Phos binding reagent is a dinuclear metal (Zn2+ or Mg2+) complex. Phos binding reagent acrylamide (PBR-A) is a specific reagent for separation of phosphorylated proteins using SDS-PAGE method. There are other sizes available. Please inqury us for quote.
Catalog No. NC1418011