Learn More
Hach Company Phosphorus (Reactive) TNTplus Vial Test (5-90 mg/L PO₄), 25 Tests
Catalog No. 501998138 Shop All Hach Company Products
Encompass
Phosphorus TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Reactive (ortho) Phosphorus (phosphate) by the Molybdovanadate method.
Catalog No. 50-199-8138 Supplier Hach Company Supplier No. TNT846
Phosphorus TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Reactive (ortho) Phosphorus (phosphate) by the Molybdovanadate method.