Learn More
Meso Scale Discovery Phos Tyr694TTL STAT5ab WCL 5
SDP
Catalog No. 5000078230
Encompass
The Meso Scale Discovery Phospho-Tyr694/Total STAT5A/B Whole Cell Lysate Kit (K15163D) is a kit for detecting phosphorylated STAT5A/B and total STAT5A/B in cell lysates The kit contains a 96-well plate coated with capture antibodies against STAT5A/B and phospho-STAT5A/B Tyr694 It also includes detection antibodies and buffers needed to perform the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range This kit can be used to monitor STAT5A/B phosphorylation and activation in response to cytokines growth factors and drugs that modulate the JAK/STAT pathway It requires a low amount of sample and provides high-quality data
Catalog No. 50-000-78230 Supplier Meso Scale Discovery Supplier No. K15163D2
The Meso Scale Discovery Phospho-Tyr694/Total STAT5A/B Whole Cell Lysate Kit (K15163D) is a kit for detecting phosphorylated STAT5A/B and total STAT5A/B in cell lysates The kit contains a 96-well plate coated with capture antibodies against STAT5A/B and phospho-STAT5A/B Tyr694 It also includes detection antibodies and buffers needed to perform the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range This kit can be used to monitor STAT5A/B phosphorylation and activation in response to cytokines growth factors and drugs that modulate the JAK/STAT pathway It requires a low amount of sample and provides high-quality data