Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosgene Solution

Catalog No. 748684 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-868-4 4 x 25 mL
74-868-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 74-868-4 Supplier Sigma Aldrich Supplier No. 7486844X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosgene is formed during the free radical induced degradation of chlorinated hydrocarbons. Determination of phosgene in air samples using thermal desorption (TD) followed by gas chromatography-mass spectrometry (GC-MS) has been reported.Phosgene solution may be used in the synthesis of:sulfonyl chloridesFmoc (Fluorenylmethyloxycarbonyl chloride)-taurylsulfonyl chlorideO-benzyl-L-serine carboxyanhydridesSure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 75-44-5
Density 0.924 g/mL at 25°C
Molecular Formula CCl2O
Linear Formula COCl2
MDL Number MFCD00036119
Quantity 4 x 25 mL
Molecular Weight (g/mol) 98.92 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.