Learn More
Hach Company Phosphorus reactive Vial Test, ULR (0.01 - 0.50 mg/L PO₄-P), 20 Tests
Catalog No. 501997625 Shop All Hach Company Products
Encompass
Phosphorous HPT chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For determination of very low phosphorous (reactive) concentrations, e.g. in wastewater effluents, surface water or drinking water samples by the Phosphormolybdenum Blue Method. Convenient handling using 50 mm rectangular vials.20 ULR Phosphorus reactive vials per box.
Catalog No. 50-199-7625 Supplier Hach Company Supplier No. HPT487
Phosphorous HPT chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For determination of very low phosphorous (reactive) concentrations, e.g. in wastewater effluents, surface water or drinking water samples by the Phosphormolybdenum Blue Method. Convenient handling using 50 mm rectangular vials.20 ULR Phosphorus reactive vials per box.