Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Phospate Buffer Giemsa/May Grunwald 6.8pH
SDP

Catalog No. 22000022
Change view
Click to view available options
Gallon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-000-022 Gallon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-022 Supplier StatLab Supplier No. BUP1057
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Phospate Buffer Giemsa/May Grunwald 6.8pH

  • Phospate Buffer Giemsa/May Grunwald 6.8pH
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
Color Clear Colorless
pH 6.8
Quantity Gallon
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.