Learn More
Apexbio Technology LLC Phosphatase Inhibitor Cocktail (2 Tubes, 100X) 100x(1ml A + 1ml B)
SDP
Catalog No. 5000064308
Encompass_Preferred
Protein phosphorylation regulates various cellular processes by controlling protein functions localization and interactions To study phosphorylation protection from endogenous phosphatases is needed as they reverse phosphorylation during sample preparation The Phosphatase Inhibitor Cocktail contains two formulations Tube A (in DMSO) targets serine/threonine phosphatases (PP1 PP2A) and alkaline phosphatases using inhibitors like Cantharidin Bromotetramisole and Microcystin LR Tube B (in aqueous solution) inhibits tyrosine acid and alkaline phosphatases using Sodium orthovanadate Sodium molybdate Sodium tartrate Imidazole and Sodium Fluoride This cocktail helps preserve phosphorylation during cell lysis and processing for biochemical assays such as immunoblotting immunoprecipitation kinase activity analysis and mass spectrometry
Catalog No. 50-000-64308 Supplier Apexbio Technology LLC Supplier No. K1015100
Protein phosphorylation regulates various cellular processes by controlling protein functions localization and interactions To study phosphorylation protection from endogenous phosphatases is needed as they reverse phosphorylation during sample preparation The Phosphatase Inhibitor Cocktail contains two formulations Tube A (in DMSO) targets serine/threonine phosphatases (PP1 PP2A) and alkaline phosphatases using inhibitors like Cantharidin Bromotetramisole and Microcystin LR Tube B (in aqueous solution) inhibits tyrosine acid and alkaline phosphatases using Sodium orthovanadate Sodium molybdate Sodium tartrate Imidazole and Sodium Fluoride This cocktail helps preserve phosphorylation during cell lysis and processing for biochemical assays such as immunoblotting immunoprecipitation kinase activity analysis and mass spectrometry