Learn More
Life Tein LLC Phospha Oseltamivir Biotin Conjugate, 1 mg, Solid powder, Purity >90%, Low water solubility, Soluble in DMSO, Undecaethylene glycol spacer
SDP
Catalog No. NC3591119
Encompass_Preferred
Phospha Oseltamivir Biotin Conjugate (1 mg) is a specialized molecular tool designed for advanced research and drug development related to the influenza virus. It combines phosphate-oseltamivir, a potent influenza neuraminidase (NA) inhibitor, with d-biotin, facilitating its application in diagnostic assays and vaccine development. The compound's robust stability on streptavidin-coated biosensors ensures its efficacy in selective influenza virus immobilization, providing critical insights into viral activity and resistance mechanisms.
  • Retains strong inhibitory action against influenza neuraminidase
  • Facilitates interaction with avidin or streptavidin detection systems
  • Low water solubility; soluble in DMSO or DMF
  • Very stable on streptavidin-coated biosensors
  • Useful in influenza diagnosis and drug discovery
Catalog No. NC3591119 Supplier Life Tein LLC Supplier No. LTF002
Add to Cart
Add to Cart
Request a Quote
Phospha Oseltamivir Biotin Conjugate (1 mg) is a specialized molecular tool designed for advanced research and drug development related to the influenza virus. It combines phosphate-oseltamivir, a potent influenza neuraminidase (NA) inhibitor, with d-biotin, facilitating its application in diagnostic assays and vaccine development. The compound's robust stability on streptavidin-coated biosensors ensures its efficacy in selective influenza virus immobilization, providing critical insights into viral activity and resistance mechanisms.
  • Retains strong inhibitory action against influenza neuraminidase
  • Facilitates interaction with avidin or streptavidin detection systems
  • Low water solubility; soluble in DMSO or DMF
  • Very stable on streptavidin-coated biosensors
  • Useful in influenza diagnosis and drug discovery