Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphatase Acid from pota
Catalog No. 502907714
Encompass
Acid phosphatase from potato is a phosphomonoesterase which can appear in multiple molecular forms of similar molecular mass but with different isoelectric points.
Catalog No. 50-290-7714 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P11461KU
Acid phosphatase from potato is a phosphomonoesterase which can appear in multiple molecular forms of similar molecular mass but with different isoelectric points.