Learn More
Worthington Biochemical Corporation PHOSPHATASE ALKALINE 250MG
SDP
Catalog No. 50592463
Encompass
Partially purified. A dried powder. Used in the NF/USP dexamethasone phosphate assay. 9001-78-9
Catalog No. 50-592-463 Supplier Worthington Biochemical Corporation Supplier No. LS003172
Partially purified. A dried powder. Used in the NF/USP dexamethasone phosphate assay. 9001-78-9