Learn More
Seracare Life Sciences Inc PHOSPHATASE ASSAY BUFFER (10X)
Catalog No. 5067413
Encompass_Preferred
Buffer for use with alkaline phosphatase based detection systems on membranes. It improves assay performance by elevating the pH for optimal activity of the phosphatase enzyme prior to addition of substrate. Intended for use with Phosphatase Wash Solution and CDP-Star chemiluminescent substrate in procedures such as Western, Southern, Northern, and dot blotting.
This item is not returnable. View return policy
Catalog No. 50-674-13 Supplier Seracare Life Sciences Inc Supplier No. 59600017
Buffer for use with alkaline phosphatase based detection systems on membranes. It improves assay performance by elevating the pH for optimal activity of the phosphatase enzyme prior to addition of substrate. Intended for use with Phosphatase Wash Solution and CDP-Star chemiluminescent substrate in procedures such as Western, Southern, Northern, and dot blotting.