Learn More
Apexbio Technology LLC Phosphatase Inhibitor Cocktail 2 (100X in ddH2O) 100x1ml
SDP
Catalog No. 503107440
Encompass_Preferred
Many cellular processes are regulated by reversible phosphorylation of proteins and approximately 30% of proteins may become phosphorylated during their lifetime Cellular crude extracts contain numerous endogenous enzymes such as phosphatases and proteases which can degrade and modify proteins present in the extract Addition of inhibitors for these enzymes is the optimal approach for maximizing intact protein yield Phosphatase Inhibitor Cocktail 2 inhibits tyrosine protein phosphatases acid phosphatases and alkaline phosphatases Phosphatase Inhibitor Cocktail 2 has been optimized and tested in cell extracts from multiple animal tissues This Phosphatase Inhibitor Cocktail 2 contains Sodium orthovanadate Sodium molybdate Sodium tartrate Imidazole and Sodium Fluoride It is supplied as a ready-to-use solution in ddH2O
Catalog No. 50-310-7440 Supplier Apexbio Technology LLC Supplier No. K1013100
Many cellular processes are regulated by reversible phosphorylation of proteins and approximately 30% of proteins may become phosphorylated during their lifetime Cellular crude extracts contain numerous endogenous enzymes such as phosphatases and proteases which can degrade and modify proteins present in the extract Addition of inhibitors for these enzymes is the optimal approach for maximizing intact protein yield Phosphatase Inhibitor Cocktail 2 inhibits tyrosine protein phosphatases acid phosphatases and alkaline phosphatases Phosphatase Inhibitor Cocktail 2 has been optimized and tested in cell extracts from multiple animal tissues This Phosphatase Inhibitor Cocktail 2 contains Sodium orthovanadate Sodium molybdate Sodium tartrate Imidazole and Sodium Fluoride It is supplied as a ready-to-use solution in ddH2O