Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Phosphatase Inhibitor Cocktail II, 1 Vial
SDP

Catalog No. 50488966
Encompass_Preferred

Lyophillized blend of five phosphatase inhibitors for use with animal tissue. A431 and Jurkat cell extracts. Suitable for extracts containing detergents. Reconstitute with 1ml water to produce a 100X concentrate. Dilute 1:100 before using.

Contains:

Imidazole
Sodium Fluoride
Sodium Molybdate
Sodium Orthovanadate
Sodium Tartrate, dihydrate

Specificity:
Protein Tyrosine Phoshatase
Acid Phosphatases
Alkaline Phosphatases

Catalog No. 50-488-966 Supplier Research Products International Corp Supplier No. P521021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lyophillized blend of five phosphatase inhibitors for use with animal tissue. A431 and Jurkat cell extracts. Suitable for extracts containing detergents. Reconstitute with 1ml water to produce a 100X concentrate. Dilute 1:100 before using.

Contains:

Imidazole
Sodium Fluoride
Sodium Molybdate
Sodium Orthovanadate
Sodium Tartrate, dihydrate

Specificity:
Protein Tyrosine Phoshatase
Acid Phosphatases
Alkaline Phosphatases

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.