Learn More
Research Products International Corp Phosphatase Inhibitor Cocktail IV, 1 Vial
SDP
Catalog No. 50488968
Encompass_Preferred
Mixture of three phosphatase inhibitors for use with animal tissue extracts. Supplied in 1ml DMSO. Dilute 1:50 before using.
Contains: (-)-p-Bromotetramisole oxalate, Cantharidin, Calyculin A, Discodermia calyx
Catalog No. 50-488-968 Supplier Research Products International Corp Supplier No. P521061
Mixture of three phosphatase inhibitors for use with animal tissue extracts. Supplied in 1ml DMSO. Dilute 1:50 before using.
Contains: (-)-p-Bromotetramisole oxalate, Cantharidin, Calyculin A, Discodermia calyx