Learn More
AG Scientific Inc Phosphatase Inhib Cocktail III
Supplier:  AG Scientific Inc P1549VIAL
Encompass_Preferred
"Phosphatase Inhibitor Cocktail III is a mixture of four phosphatase inhibitors which has been optimized for the inhibition of both serine/threonine and protein tyrosine phosphatases. Reconstitute each vial with 1 mL water to obtain a 1 mL stock solution. Each vial contains: 250 mM Sodium fluoride 50 mM Sodium orthovanadate 50 mM Sodium pyrophosphate 50 mM β-glycerophosphate"
Catalog No. 50-252-2600