Learn More
AG Scientific Inc Phosphatase Inhib I
Catalog No. 502522609
Encompass_Preferred
"Phosphatase Inhibitor Cocktail I is a specially formulated mixture of three phosphatase inhibitors that will inhibit alkaline phosphatases and serine/threonine protein phosphatases such as PP1 and PP2A. A specially formulated cocktail of three phosphatase inhibitors that will inhibit alkaline phosphatases and serine/threonine protein phosphatases such as PP1 and PP2A. Recommended for use with tissue and cell extracts, including extracts containing detergents. Each vial contains: 2.5 mM (-)-p-bromotetramisole oxalate 500 μM Cantharidin 500 nM microcystin-LR"
Catalog No. 50-252-2609 Supplier AG Scientific Inc Supplier No. P1517VIAL
"Phosphatase Inhibitor Cocktail I is a specially formulated mixture of three phosphatase inhibitors that will inhibit alkaline phosphatases and serine/threonine protein phosphatases such as PP1 and PP2A. A specially formulated cocktail of three phosphatase inhibitors that will inhibit alkaline phosphatases and serine/threonine protein phosphatases such as PP1 and PP2A. Recommended for use with tissue and cell extracts, including extracts containing detergents. Each vial contains: 2.5 mM (-)-p-bromotetramisole oxalate 500 μM Cantharidin 500 nM microcystin-LR"