Learn More
Enzo Life Sciences SCREEN-WELL Phosphatase Inhibitor library (500 µl/well)
SDP
Supplier:  Enzo Life Sciences BML28340500
Encompass_Preferred
The SCREEN-WELL Phosphatase Inhibitor Library contains 33 known phosphatase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 µl or 500 µl per well. The library is ideal for chemical genomics, assay development, and as a reference set for secondary screening. Includes inhibitors of these important phosphatases: Calcineurin(PP2B), JSP-1, PRL-1, CD45, PP1, PRL-3, CDC25, PP2A, PTEN, and many more. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0211