Learn More
Santa Cruz Biotechnology Phosphatase Inhibitor Cocktail A, 1 mL
Catalog No. NC9529581
Encompass
Phosphatase Inhibitor Cocktail A is an inhibitor of L-isozymes of alkaline phosphatase and ser/thr protein phosphatases.
Catalog No. NC9529581 Supplier Santa Cruz Biotechnology Supplier No. SC45044
Add to Cart
Add to Cart
Phosphatase Inhibitor Cocktail A is an inhibitor of L-isozymes of alkaline phosphatase and ser/thr protein phosphatases.