Learn More
Sigma Aldrich Fine Chemicals Biosciences PhosSTOP, 20 tablets
Catalog No. NC0927101
Encompass
PhosSTOP tablets are a proprietary blend of phosphatase inhibitors, formulated as a ready-to-use tablet which dissolves quickly in aqueous solutions (buffers), including buffers containing formalin (for formalin fixation).
Catalog No. NC0927101 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4906837001
PhosSTOP tablets are a proprietary blend of phosphatase inhibitors, formulated as a ready-to-use tablet which dissolves quickly in aqueous solutions (buffers), including buffers containing formalin (for formalin fixation).