Learn More
Apexbio Technology LLC Phosphatase Inhibitor Cocktail 2 (100X in ddH2O), 1ml x 10
SDP
Catalog No. 501905551
Encompass_Preferred
For tyrosine protein phosphatases, acid and alkaline phosphatases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5551 Supplier Apexbio Technology LLC Supplier No. K101310
For tyrosine protein phosphatases, acid and alkaline phosphatases. Other sizes are available. Please inqury us for quote.