Learn More
Apexbio Technology LLC Phosphatase Inhibitor Cocktail 1 (100X in DMSO), 1ml x 50
SDP
Catalog No. 501905549
Encompass_Preferred
For serine/threonine protein phosphatases and L-isozymes of alkaline phosphatases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5549 Supplier Apexbio Technology LLC Supplier No. K101250
For serine/threonine protein phosphatases and L-isozymes of alkaline phosphatases. Other sizes are available. Please inqury us for quote.