Learn More
Medchemexpress LLC Phosphatase Inhibitor Cocktail III (100x in DMSO) | 1 ML
SDP
Catalog No. 5000366056
Encompass_Preferred
Phosphatase Inhibitor Cocktail III (100x in DMSO) is a blend of three phosphatase inhibitors designed to effectively inhibit serine/threonine phosphatases and alkaline phosphatases. This cocktail helps maintain the phosphorylation status of proteins by preventing degradation, which is often enhanced when cells are studied in vitro.
  • Effectively inhibits serine/threonine phosphatases
  • Effectively inhibits alkaline phosphatases
  • Maintains the phosphorylation status of proteins
  • Prevents protein degradation
  • Suitable for Western Blot analysis
  • Suitable for IP
  • Suitable for Co-IP
  • Suitable for pull-down assays
  • Suitable for immunofluorescence (IF)
  • Suitable for immunohistochemistry (IHC)
  • Suitable for kinase assays
Catalog No. 50-003-66056 Supplier Medchemexpress LLC Supplier No. 1MLHYK0023
Phosphatase Inhibitor Cocktail III (100x in DMSO) is a blend of three phosphatase inhibitors designed to effectively inhibit serine/threonine phosphatases and alkaline phosphatases. This cocktail helps maintain the phosphorylation status of proteins by preventing degradation, which is often enhanced when cells are studied in vitro.
  • Effectively inhibits serine/threonine phosphatases
  • Effectively inhibits alkaline phosphatases
  • Maintains the phosphorylation status of proteins
  • Prevents protein degradation
  • Suitable for Western Blot analysis
  • Suitable for IP
  • Suitable for Co-IP
  • Suitable for pull-down assays
  • Suitable for immunofluorescence (IF)
  • Suitable for immunohistochemistry (IHC)
  • Suitable for kinase assays