Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Phosphatase Inhibitor Cocktail I, 1 Vial
SDP

Catalog No. 50488965
Encompass_Preferred

Mixture of three phosphatase inhibitors for use with animal tissue. A431 and Jurkat cell extracts. Suitable for extracts containing detergents. Supplied in 1ml DMSO. Dilute 1:100 before using.

Contains:

(-)-p-Bromotetramisole oxalate
Cantharidin
Microcystin LR, Microcystis, Aeruginosa

Specificity:
Alkaline Phosphatases and Ser/Thr
Phosphatases 1 and 2A (PP-l and PP-2A)

Catalog No. 50-488-965 Supplier Research Products International Corp Supplier No. P521001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mixture of three phosphatase inhibitors for use with animal tissue. A431 and Jurkat cell extracts. Suitable for extracts containing detergents. Supplied in 1ml DMSO. Dilute 1:100 before using.

Contains:

(-)-p-Bromotetramisole oxalate
Cantharidin
Microcystin LR, Microcystis, Aeruginosa

Specificity:
Alkaline Phosphatases and Ser/Thr
Phosphatases 1 and 2A (PP-l and PP-2A)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.