Learn More
Seracare Life Sciences Inc PHOSPHATASE WASH SOLUTION (5X)
Supplier:  Seracare Life Sciences Inc 59600018
Encompass_Preferred
Wash solution for use with alkaline phosphatase based detection systems on membranes. It is formulated to effectively remove unbound phosphatase conjugate and improve assay performance. Phosphatase Wash Solution is intended for use with Phosphatase Assay Buffer and CDP-Star Chemiluminescent Substrate in procedures such as Western, Southern, Northern and dot blotting.
Catalog No. 50-674-14