Learn More
SPEX CERTIPREP LLC 3P PHOSPHATE 250ML 1000PPM
Catalog No. NC9636385
Encompass_Preferred
(HPS by Zeptometrix) 1000 ug/mL Phosphate in H2O; Single component IC Standard, 18 month expiry date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9636385 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICPPM250
(HPS by Zeptometrix) 1000 ug/mL Phosphate in H2O; Single component IC Standard, 18 month expiry date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.