Learn More
Hach Company Phosphate Standard Solution, 50 mg/L as PO₄ (NIST), pk/20 - 2 mL Ampules
Catalog No. 501997385 Shop All Hach Company Products
Encompass
For accuracy checks in phosphate (phosphorus) determinations
Catalog No. 50-199-7385 Supplier Hach Company Supplier No. 17120H
For accuracy checks in phosphate (phosphorus) determinations