Learn More
Roche Diagnostics Phosphate Bio, 200 tests
Supplier:  Roche Diagnostics 06990070001
Encompass
Phosphate is a mineral nutrient, which is reqired in cell cultures and microbial fermentation for optimal growth and product formation. For control of culture media and for monitoring in a culture process, the phosphate concentration can be determined on a Cedex Bio or Cedex Bio HT Analyzer with high reliability and convenience.
Catalog No. NC1351981