Learn More
Roche Diagnostics Metabolite testing reagent set
Supplier:  Roche Diagnostics 06990088001
Encompass
Metabolite testing reagent set for quantitative determination of Phosphate in aqueous solution or fermentation broth using the Cedex Bio HT instrument.
Catalog No. 50-589-448