Learn More
General Laboratory Products Phosphate Buffered Water, 9mL 100/pk
Catalog No. 50371475
Encompass_Preferred
Phosphate Buffered Water - 9ml, 16x125mm polycarbonate tube, 100 per case. For use as a diluent in microbial limit testing of food and water. 
Catalog No. 50371475 Supplier General Laboratory Products Supplier No. 16T28009
Phosphate Buffered Water - 9ml, 16x125mm polycarbonate tube, 100 per case. For use as a diluent in microbial limit testing of food and water. 