Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer, 0.05 M, pH 5.80, Ricca Chemical
SDP

Catalog No. R579400020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R579400020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R579400020F Supplier Ricca Chemical Company Supplier No. R579400020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use phosphate buffer solution formulated for pH control.

Specifications

Chemical Name or Material Phosphate Buffer
CAS 7732-18-5, 7778-77-0, 1310-73-2
CAS Max % 99.31, 0.68, 0.01
Color Colorless Liquid
Concentration 0.05 M
For Use With (Application) Water Analysis
pH 5.8
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Sterility Non-sterile
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.