Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer, 0.10 M, pH 6.20, Ricca Chemical
SDP

Catalog No. 579925 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5799-25 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5799-25 Supplier Ricca Chemical Company Supplier No. 5799.25
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphate buffer for general laboratory use.

Specifications

Chemical Name or Material Phosphate Buffer
CAS 7732-18-5, 10049-21-5, 1310-73-2
CAS Min % 96.0, 1.33, 0.21
CAS Max % 100.0, 1.39, 0.22
Color Colorless
Concentration 0.1 M
For Use With (Application) Water Analysis
pH 6
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.10 Molar, pH 6.20
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.