Learn More
Electron Microscopy Sciences Phosphate Buffer 0.1%, pH 6.8-7.0 500 ML
SDP
Catalog No. 5032133
Encompass_Preferred
Used in Maximow's Method for Bone Marrow, Morphology details of marrow cells. Mallory (1961). Morphology details of marrow cells. See AFIP Manual of Histologic Staining Methods, 3rd Ed, Ed. L. Luna: New York: McGraw Hill Publications, c. 1968, p. 120 or Mallory, F.B., Pathological technique, New York, Hafner Publishing Co., 1961, p.196
Catalog No. 50-321-33 Supplier Electron Microscopy Sciences Supplier No. 2625204
Used in Maximow's Method for Bone Marrow, Morphology details of marrow cells. Mallory (1961). Morphology details of marrow cells. See AFIP Manual of Histologic Staining Methods, 3rd Ed, Ed. L. Luna: New York: McGraw Hill Publications, c. 1968, p. 120 or Mallory, F.B., Pathological technique, New York, Hafner Publishing Co., 1961, p.196