Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioworld PHOSPHATE BUFFER 1M, PH 8.5 1L

Catalog No. NC1948009
Encompass_Preferred

Phosphate Buffer 1M, pH 8.5, Endotoxin Free, 1 L

Catalog No. NC1948009 Supplier Bioworld Supplier No. 401212761
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphate Buffer 1M, pH 8.5, Endotoxin Free, 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.