Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS Phosphate Buffer

Catalog No. 09316763 Shop All GVS Products
Change view
Click to view available options
Fill Volume:
99 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
09-316-763 99 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-316-763 Supplier GVS Supplier No. 10498503S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains monobasic potassium phosphate and deionized water

  • Pre-filled sterile dilution bottle containing Butterfield's Phosphate Buffer

Specifications

Form Solution
Manufacturing Origin China
Product Type Broth
Sterility Non-sterile
Target Organism Class Bacteria
Test Type Microbiological Tests
Fill Volume 99 mL
Quantity 72/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.