Learn More
Nelson Jameson Company 3M Phosphate Buffer 90ml
Catalog No. NC1797200
Encompass
3M™ Flip-Top Dilution Bottles are ideal diluents for enumeration of microoroganism from food, beverage and environmental samples. The solution is decontaminated through gamma irradiation for long shelf life, and the updated handy flip-top style provides a fast, easy and reliable for sample processing
Catalog No. NC1797200 Supplier Nelson Jameson Company Supplier No. 3513603 Y
3M™ Flip-Top Dilution Bottles are ideal diluents for enumeration of microoroganism from food, beverage and environmental samples. The solution is decontaminated through gamma irradiation for long shelf life, and the updated handy flip-top style provides a fast, easy and reliable for sample processing