Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphate Buffer, for Cyanide, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffer |
| CAS | 10049-21-5 |
| Concentration | 1X |
| For Use With (Application) | Water Analysis |
| pH | 4 to 5 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Packaging | Natural Poly Bottle |
| Grade | Laboratory |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction