Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer, for Cyanide, Ricca Chemical
SDP

Catalog No. R581032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R581032 1 L
5810-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R581032 Supplier Ricca Chemical Company Supplier No. 581032
Add to Cart
Add to Cart

For the Analysis of Cyanide and Cyanogen Chloride

Specifications

Chemical Name or Material Phosphate Buffer
Name Note For Cyanide
CAS 10049-21-5, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Water Analysis
pH 4 to 5
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.